siRNA Details
| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145877.1 | NP_665884.1 | 4 | 398 | ||
| GenBank mRNA | AJ000696.1 | - | 4 | 398 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif1c| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145877.1 | NP_665884.1 | 4 | 398 | ||
| GenBank mRNA | AJ000696.1 | - | 4 | 398 | ||