siRNA Details
| Gene Symbol: Fabp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145878.1 | NP_665885.1 | 1, 2 | 121 | ||
| GenBank mRNA | FQ210725.1 | - | 1, 2 | 131 | ||
| FQ228381.1 | - | 1, 2 | 133 | |||
| S69874.1 | - | 1, 2 | 123 | |||
| S83247.1 | - | 1, 2 | 138 | |||
| U13253.1 | - | 1, 2 | 121 | |||

Fabp5
Genomic Map