siRNA Details
| Gene Symbol: Zwint | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147138.2 | NP_671479.1 | 2, 3 | 212 | ||
| GenBank mRNA | AF439397.1 | - | 2, 3 | 194 | ||
| AF532776.1 | - | 2, 3 | 212 | |||
| BC063144.1 | - | 2, 3 | 204 | 6921645 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zwint| Gene Symbol: Zwint | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147138.2 | NP_671479.1 | 2, 3 | 212 | ||
| GenBank mRNA | AF439397.1 | - | 2, 3 | 194 | ||
| AF532776.1 | - | 2, 3 | 212 | |||
| BC063144.1 | - | 2, 3 | 204 | 6921645 | ||