siRNA Details
| Gene Symbol: Dclre1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147145.2 | NP_671486.2 | 1, 2 | 139 | ||
| GenBank mRNA | AF395746.1 | - | 1, 2 | 171 | ||
| BC083546.1 | - | 1, 2 | 139 | 7134940 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dclre1c| Gene Symbol: Dclre1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147145.2 | NP_671486.2 | 1, 2 | 139 | ||
| GenBank mRNA | AF395746.1 | - | 1, 2 | 171 | ||
| BC083546.1 | - | 1, 2 | 139 | 7134940 | ||