siRNA Details
| Gene Symbol: Ostf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148892.3 | NP_683690.2 | 4 | 315 | ||
| XM_008760287.1 | XP_008758509.1 | 4 | 310 | |||
| GenBank mRNA | AF523266.1 | - | 4 | 194 | ||
| BC062400.1 | - | 4 | 294 | 5621572 | ||
| FM048871.1 | - | 4 | 315 | |||

Ostf1
Genomic Map