siRNA Details
| Gene Symbol: Trib3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144755.2 | NP_653356.1 | 4 | 1150 | ||
| GenBank mRNA | AB020967.1 | - | 1135 | |||
| BC091120.1 | - | 4 | 1150 | 7326024 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trib3| Gene Symbol: Trib3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144755.2 | NP_653356.1 | 4 | 1150 | ||
| GenBank mRNA | AB020967.1 | - | 1135 | |||
| BC091120.1 | - | 4 | 1150 | 7326024 | ||