siRNA Details
| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145676.1 | NP_663709.1 | 5 | 438 | ||
| XM_006241967.2 | XP_006242029.1 | 6 | 555 | |||
| GenBank mRNA | AF393211.1 | - | 5 | 438 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cby1| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145676.1 | NP_663709.1 | 5 | 438 | ||
| XM_006241967.2 | XP_006242029.1 | 6 | 555 | |||
| GenBank mRNA | AF393211.1 | - | 5 | 438 | ||