siRNA Details
| Gene Symbol: Slc25a25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145677.1 | NP_663710.1 | 2 | 291 | ||
| XM_006233931.2 | XP_006233993.1 | 2 | 412 | |||
| XM_006233932.2 | XP_006233994.1 | 2 | 412 | |||
| XM_006233933.2 | XP_006233995.1 | 2 | 1064 | |||
| XM_006233934.2 | XP_006233996.1 | 2 | 1064 | |||
| XM_006233935.2 | XP_006233997.1 | 2 | 1628 | |||
| XM_006233937.2 | XP_006233999.1 | 2 | 648 | |||
| GenBank mRNA | AY043169.1 | - | 2 | 291 | ||
| FQ211394.1 | - | 2 | 307 | |||

Slc25a25
Genomic Map