siRNA Details
| Gene Symbol: Vps4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145678.1 | NP_663711.1 | 3 | 167 | ||
| XM_006255560.1 | XP_006255622.1 | 3 | 386 | |||
| GenBank mRNA | AB076398.1 | - | 3 | 167 | ||
| BC070931.1 | - | 3 | 270 | 7096867 | ||
| FQ213218.1 | - | 347 | ||||

Vps4a
Genomic Map