siRNA Details
| Gene Symbol: Tnfsf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145681.1 | NP_663714.1 | 2 | 167 | ||
| GenBank mRNA | AY115578.1 | - | 2 | 167 | ||
| EF030546.1 | - | 2 | 158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfsf10| Gene Symbol: Tnfsf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145681.1 | NP_663714.1 | 2 | 167 | ||
| GenBank mRNA | AY115578.1 | - | 2 | 167 | ||
| EF030546.1 | - | 2 | 158 | |||