siRNA Details
Gene Symbol: LOC252831 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145722.1 | NP_663774.1 | 915 | |||
GenBank mRNA | AB077475.1 | - | 915 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC252831 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145722.1 | NP_663774.1 | 915 | |||
GenBank mRNA | AB077475.1 | - | 915 |