siRNA Details
| Gene Symbol: Pzp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145779.2 | NP_665722.2 | 2 | 201 | ||
| XM_008763310.1 | XP_008761532.1 | 2 | 212 | |||
| GenBank mRNA | FQ209918.1 | - | 2 | 230 | ||
| FQ210288.1 | - | 2 | 230 | |||
| M77183.1 | - | 2 | 197 | |||
| M84000.1 | - | 2 | 240 | |||

Pzp
Genomic Map