siRNA Details
| Gene Symbol: Hdgfrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145785.1 | NP_665728.1 | 4 | 500 | ||
| XM_008759496.1 | XP_008757718.1 | 4 | 775 | |||
| GenBank mRNA | AF389347.1 | - | 4 | 500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hdgfrp3| Gene Symbol: Hdgfrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145785.1 | NP_665728.1 | 4 | 500 | ||
| XM_008759496.1 | XP_008757718.1 | 4 | 775 | |||
| GenBank mRNA | AF389347.1 | - | 4 | 500 | ||