siRNA Details
| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145877.1 | NP_665884.1 | 5 | 453 | ||
| XM_006246564.2 | XP_006246626.1 | 5 | 527 | |||
| XM_006246565.2 | XP_006246627.1 | 4 | 407 | |||
| GenBank mRNA | AJ000696.1 | - | 5 | 453 | ||

Kif1c
Genomic Map