siRNA Details
| Gene Symbol: Slc35a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147140.2 | NP_671481.2 | 2 | 1480 | ||
| XM_006254528.1 | XP_006254590.1 | 2 | 1514 | |||
| XM_006254529.2 | XP_006254591.1 | 4 | 1346 | |||
| XM_008772019.1 | XP_008770241.1 | 2 | 1433 | |||
| GenBank mRNA | AF406814.1 | - | 1 | 1251 | ||
| BC085695.1 | - | 2 | 1480 | 7191954 | ||
| BC086986.1 | - | 2 | 1433 | 7192444 | ||
| FQ224389.1 | - | 4 | 1351 | |||

Slc35a4
Genomic Map