siRNA Details
| Gene Symbol: Nr1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147210.2 | NP_671743.2 | 3, 4 | 580 | ||
| GenBank mRNA | U15660.1 | - | 2, 3 | 297 | ||
| U20796.1 | - | 2, 3 | 237 | |||
| X78135.1 | - | 3, 4 | 433 | |||
| X82777.1 | - | 3, 4 | 515 | |||

Nr1d2
Genomic Map