siRNA Details
| Gene Symbol: Pcyox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145085.2 | NP_659553.1 | 6 | 1047 | ||
| GenBank mRNA | AF332142.1 | - | 1037 | |||
| BC078719.1 | - | 6 | 1047 | 7127920 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcyox1| Gene Symbol: Pcyox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145085.2 | NP_659553.1 | 6 | 1047 | ||
| GenBank mRNA | AF332142.1 | - | 1037 | |||
| BC078719.1 | - | 6 | 1047 | 7127920 | ||