siRNA Details
| Gene Symbol: Sbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147135.2 | NP_671476.1 | 3 | 865 | ||
| XM_008759853.1 | XP_008758075.1 | 3 | 872 | |||
| XM_008759854.1 | XP_008758076.1 | 4 | 1200 | |||
| GenBank mRNA | AB010154.1 | - | 3 | 577 | ||
| BC131843.1 | - | 3 | 865 | 8363395 | ||
| Gene Symbol: LOC103690016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758862.1 | XP_008757084.1 | 3 | 913 | ||
| XM_008758863.1 | XP_008757085.1 | 3 | 870 | |||
| XM_008758864.1 | XP_008757086.1 | 4 | 1174 | |||

Sbk1
Genomic Map