siRNA Details
| Gene Symbol: Ruvbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147177.1 | NP_671706.1 | 881 | |||
| XM_006236865.2 | XP_006236927.1 | 955 | ||||
| GenBank mRNA | AB001581.1 | - | 868 | |||
| AB002406.1 | - | 881 | ||||
| BC072511.1 | - | 866 | 7104777 | |||
| BC086531.1 | - | 899 | 7304521 | |||
| Gene Symbol: RGD1564855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773046.1 | XP_008771268.1 | 1 | 926 | ||

Ruvbl1
Genomic Map