siRNA Details
| Gene Symbol: Snx27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110151.1 | NP_001103621.1 | 1, 2 | 431 | ||
| NM_152847.2 | NP_690060.2 | 1, 2 | 431 | |||
| GenBank mRNA | AB010245.1 | - | 1, 2 | 431 | ||
| AB051816.1 | - | 1, 2 | 431 | |||
| BF524912.1 | - | 1, 2 | 324 | |||

Snx27
Genomic Map