siRNA Details
| Gene Symbol: Rab8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153317.1 | NP_695229.1 | 3 | 275 | ||
| GenBank mRNA | BC158579.1 | - | 3 | 301 | 9091689 | |
| U53475.1 | - | 3 | 275 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab8b| Gene Symbol: Rab8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153317.1 | NP_695229.1 | 3 | 275 | ||
| GenBank mRNA | BC158579.1 | - | 3 | 301 | 9091689 | |
| U53475.1 | - | 3 | 275 | |||