siRNA Details
Gene Symbol: Krt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153476.1 | NP_703206.1 | 1 | 297 | ||
GenBank mRNA | AY128946.1 | - | 1 | 297 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153476.1 | NP_703206.1 | 1 | 297 | ||
GenBank mRNA | AY128946.1 | - | 1 | 297 |