siRNA Details
| Gene Symbol: Rcan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153724.2 | NP_714946.2 | 1, 2 | 156 | ||
| GenBank mRNA | AB075973.1 | - | 1, 2 | 112 | ||
| BC072478.1 | - | 1, 2 | 156 | 7103498 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rcan1| Gene Symbol: Rcan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153724.2 | NP_714946.2 | 1, 2 | 156 | ||
| GenBank mRNA | AB075973.1 | - | 1, 2 | 112 | ||
| BC072478.1 | - | 1, 2 | 156 | 7103498 | ||