siRNA Details
| Gene Symbol: Dcps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153302.1 | NP_695214.1 | 2, 3 | 370 | ||
| GenBank mRNA | AF411249.1 | - | 2, 3 | 370 | ||
| BC089773.1 | - | 2, 3 | 365 | |||
| BC105900.1 | - | 2, 3 | 375 | 7377732 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcps| Gene Symbol: Dcps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153302.1 | NP_695214.1 | 2, 3 | 370 | ||
| GenBank mRNA | AF411249.1 | - | 2, 3 | 370 | ||
| BC089773.1 | - | 2, 3 | 365 | |||
| BC105900.1 | - | 2, 3 | 375 | 7377732 | ||