siRNA Details
| Gene Symbol: Grina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153308.4 | NP_695220.4 | 3 | 537 | ||
| GenBank mRNA | BC062074.1 | - | 3 | 507 | 5599428 | |
| CK475470.1 | - | 3 | 537 | |||
| S61973.1 | - | 3 | 487 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grina| Gene Symbol: Grina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153308.4 | NP_695220.4 | 3 | 537 | ||
| GenBank mRNA | BC062074.1 | - | 3 | 507 | 5599428 | |
| CK475470.1 | - | 3 | 537 | |||
| S61973.1 | - | 3 | 487 | |||