siRNA Details
Gene Symbol: Mina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153309.2 | NP_695221.1 | 2 | 222 | ||
XM_008768680.1 | XP_008766902.1 | 2 | 287 | |||
GenBank mRNA | AB083195.1 | - | 1 | 197 | ||
BC087650.1 | - | 2 | 222 | 7310399 |
