siRNA Details
| Gene Symbol: SIX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016932.4 | NP_058628.3 | 1, 2 | 856 | ||
| GenBank mRNA | AF136940.1 | - | 1, 2 | 615 | ||
| AF332196.1 | - | 1, 2 | 553 | |||
| AF332197.1 | - | 1, 2 | 856 | |||
| BC024033.2 | - | 1, 2 | 836 | 3027992 | ||
| BT020100.1 | - | 1, 2 | 563 | |||

SIX2
Genomic Map