siRNA Details
Gene Symbol: Lzts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153470.1 | NP_703200.1 | 3 | 825 | ||
GenBank mRNA | AB075607.1 | - | 3 | 825 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lzts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153470.1 | NP_703200.1 | 3 | 825 | ||
GenBank mRNA | AB075607.1 | - | 3 | 825 |