siRNA Details
Gene Symbol: Sec11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153628.2 | NP_705892.1 | 2 | 211 | ||
GenBank mRNA | AB022714.2 | - | 2 | 212 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sec11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153628.2 | NP_705892.1 | 2 | 211 | ||
GenBank mRNA | AB022714.2 | - | 2 | 212 |