siRNA Details
| Gene Symbol: Itgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153720.1 | NP_714942.1 | 4 | 418 | ||
| GenBank mRNA | AJ440952.1 | - | 4 | 418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itgb3| Gene Symbol: Itgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153720.1 | NP_714942.1 | 4 | 418 | ||
| GenBank mRNA | AJ440952.1 | - | 4 | 418 | ||