siRNA Details
Gene Symbol: Ache | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172009.1 | NP_742006.1 | 1 | 527 | ||
GenBank mRNA | BC094521.1 | - | 622 | 7315968 | ||
S50879.1 | - | 1 | 527 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ache | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172009.1 | NP_742006.1 | 1 | 527 | ||
GenBank mRNA | BC094521.1 | - | 622 | 7315968 | ||
S50879.1 | - | 1 | 527 |