siRNA Details
| Gene Symbol: Fgl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172010.1 | NP_742007.1 | 3 | 290 | ||
| GenBank mRNA | AF508020.1 | - | 3 | 290 | ||
| BC088107.1 | - | 3 | 246 | 7368731 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgl1| Gene Symbol: Fgl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172010.1 | NP_742007.1 | 3 | 290 | ||
| GenBank mRNA | AF508020.1 | - | 3 | 290 | ||
| BC088107.1 | - | 3 | 246 | 7368731 | ||