siRNA Details
Gene Symbol: Rbm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152861.4 | NP_690600.1 | 16 | 2020 | ||
GenBank mRNA | D83948.1 | - | 16 | 2020 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152861.4 | NP_690600.1 | 16 | 2020 | ||
GenBank mRNA | D83948.1 | - | 16 | 2020 |