siRNA Details
| Gene Symbol: Rcan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153724.2 | NP_714946.2 | 2 | 193 | ||
| XM_006248041.2 | XP_006248103.1 | 2 | 393 | |||
| GenBank mRNA | AB075973.1 | - | 2 | 149 | ||
| BC072478.1 | - | 2 | 193 | 7103498 | ||

Rcan1
Genomic Map