siRNA Details
| Gene Symbol: Cd247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205304.1 | NP_001192233.1 | 4, 5 | 371 | ||
| NM_170789.2 | NP_740770.2 | 5, 6 | 452 | |||
| XM_006250147.2 | XP_006250209.1 | 5, 6 | 492 | |||
| GenBank mRNA | BC097933.1 | - | 5, 6 | 395 | 7384006 | |
| CO562119.1 | - | 4, 5 | 305 | |||
| D13555.1 | - | 5, 6 | 452 | |||
| L08447.1 | - | 5, 6 | 379 | |||

Cd247
Genomic Map