siRNA Details
| Gene Symbol: Vps45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172072.2 | NP_742069.1 | 1 | 128 | ||
| GenBank mRNA | BC081705.1 | - | 1 | 128 | 7192410 | |
| U81160.1 | - | 1 | 128 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps45| Gene Symbol: Vps45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172072.2 | NP_742069.1 | 1 | 128 | ||
| GenBank mRNA | BC081705.1 | - | 1 | 128 | 7192410 | |
| U81160.1 | - | 1 | 128 | |||