siRNA Details
| Gene Symbol: Zfp161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172325.2 | NP_758828.2 | 186 | |||
| GenBank mRNA | AY157624.1 | - | 1 | 105 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp161| Gene Symbol: Zfp161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172325.2 | NP_758828.2 | 186 | |||
| GenBank mRNA | AY157624.1 | - | 1 | 105 | ||