siRNA Details
| Gene Symbol: Abca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173307.1 | NP_775429.1 | 2 | 123 | ||
| XM_006247560.2 | XP_006247622.1 | 2 | 324 | |||
| GenBank mRNA | AJ426052.2 | - | 2 | 123 | ||
| AJ550165.1 | - | 1 | 96 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abca5| Gene Symbol: Abca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173307.1 | NP_775429.1 | 2 | 123 | ||
| XM_006247560.2 | XP_006247622.1 | 2 | 324 | |||
| GenBank mRNA | AJ426052.2 | - | 2 | 123 | ||
| AJ550165.1 | - | 1 | 96 | |||