siRNA Details
| Gene Symbol: LOC286987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173321.1 | NP_775443.1 | 151 | |||
| GenBank mRNA | M60388.1 | - | 151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC286987| Gene Symbol: LOC286987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173321.1 | NP_775443.1 | 151 | |||
| GenBank mRNA | M60388.1 | - | 151 | |||