siRNA Details
Gene Symbol: Lgr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173328.1 | NP_775450.1 | 5 | 489 | ||
GenBank mRNA | AF061443.1 | - | 5 | 489 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lgr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173328.1 | NP_775450.1 | 5 | 489 | ||
GenBank mRNA | AF061443.1 | - | 5 | 489 |