siRNA Details
| Gene Symbol: Fntb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172034.2 | NP_742031.1 | 2 | 221 | ||
| GenBank mRNA | BC087675.1 | - | 2 | 221 | 7319293 | |
| M69056.1 | - | 2 | 235 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fntb| Gene Symbol: Fntb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172034.2 | NP_742031.1 | 2 | 221 | ||
| GenBank mRNA | BC087675.1 | - | 2 | 221 | 7319293 | |
| M69056.1 | - | 2 | 235 | |||