siRNA Details
| Gene Symbol: Eaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172047.1 | NP_742044.1 | 2 | 292 | ||
| XM_008768707.1 | XP_008766929.1 | 2 | 179 | |||
| GenBank mRNA | AY049022.1 | - | 2 | 292 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eaf2| Gene Symbol: Eaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172047.1 | NP_742044.1 | 2 | 292 | ||
| XM_008768707.1 | XP_008766929.1 | 2 | 179 | |||
| GenBank mRNA | AY049022.1 | - | 2 | 292 | ||