siRNA Details
Gene Symbol: Slc30a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172066.1 | NP_742063.1 | 3 | 878 | ||
GenBank mRNA | Y16774.1 | - | 3 | 878 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc30a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172066.1 | NP_742063.1 | 3 | 878 | ||
GenBank mRNA | Y16774.1 | - | 3 | 878 |