siRNA Details
| Gene Symbol: Hsbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173119.2 | NP_775142.1 | 3 | 270 | ||
| GenBank mRNA | AF522937.1 | - | 3 | 217 | ||
| BC058131.1 | - | 3 | 255 | 6919748 | ||
| EX491168.1 | - | 3 | 270 | |||
| FQ211891.1 | - | 3 | 238 | |||
| FQ212621.1 | - | 3 | 242 | |||
| FQ213728.1 | - | 3 | 212 | |||
| FQ216175.1 | - | 3 | 249 | |||
| FQ229155.1 | - | 3 | 255 | |||
| FQ229754.1 | - | 3 | 229 | |||
| FQ230047.1 | - | 3 | 227 | |||
| FQ230722.1 | - | 3 | 229 | |||
| FQ233773.1 | - | 3 | 228 | |||

Hsbp1
Genomic Map