siRNA Details
| Gene Symbol: Abi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173143.1 | NP_775166.1 | 2, 3 | 323 | ||
| GenBank mRNA | BC097273.1 | - | 381 | |||
| FQ222350.1 | - | 693 | ||||
| U94904.1 | - | 2, 3 | 323 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abi2| Gene Symbol: Abi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173143.1 | NP_775166.1 | 2, 3 | 323 | ||
| GenBank mRNA | BC097273.1 | - | 381 | |||
| FQ222350.1 | - | 693 | ||||
| U94904.1 | - | 2, 3 | 323 | |||