siRNA Details
| Gene Symbol: Abca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173307.1 | NP_775429.1 | 2, 3 | 145 | ||
| XM_006247560.2 | XP_006247622.1 | 2, 3 | 346 | |||
| GenBank mRNA | AJ426052.2 | - | 2, 3 | 145 | ||
| AJ550165.1 | - | 1, 2 | 118 | |||

Abca5
Genomic Map