siRNA Details
| Gene Symbol: Gcnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173312.2 | NP_775434.1 | 2 | 474 | ||
| XM_006243366.2 | XP_006243428.1 | 5 | 1169 | |||
| XM_008766220.1 | XP_008764442.1 | 4 | 681 | |||
| GenBank mRNA | AB098520.1 | - | 2 | 475 | ||

Gcnt3
Genomic Map