siRNA Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176074.3 | NP_788263.1 | 4 | 522 | ||
| XM_008760747.1 | XP_008758969.1 | 5 | 711 | |||
| XM_008760749.1 | XP_008758971.1 | 5 | 696 | |||
| XM_008760750.1 | XP_008758972.1 | 5 | 710 | |||
| GenBank mRNA | AY230250.1 | - | 4 | 524 | ||

C6
Genomic Map