siRNA Details
| Gene Symbol: P4ha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172062.2 | NP_742059.2 | 10, 11 | 1491 | ||
| XM_006256424.2 | XP_006256486.1 | 11, 12 | 1576 | |||
| XM_006256425.2 | XP_006256487.1 | 10, 11 | 1485 | |||
| GenBank mRNA | BC078703.1 | - | 10, 11 | 1491 | 7115543 | |
| X78949.1 | - | 10, 11 | 1325 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: U16162.

P4ha1
Genomic Map