siRNA Details
| Gene Symbol: Timm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172074.3 | NP_742071.1 | 2 | 798 | ||
| XM_006234473.2 | XP_006234535.1 | 3 | 410 | |||
| XM_006234474.2 | XP_006234536.1 | 4 | 522 | |||
| GenBank mRNA | AF150091.1 | - | 2 | 233 | ||
| BC091116.1 | - | 2 | 799 | 7366212 | ||

Timm10
Genomic Map